missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SGCG |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838144
|
Proteintech
18102-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828144
|
Proteintech
18102-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.Specifications
SGCG | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P82348, Q13326 | |
24053, 305941, 6445 | |
SGCG Fusion Protein Ag12402 | |
Primary | |
-20°C | |
SGCG |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
35DAG, A4, DAGA4, DMDA, DMDA1, Gamma sarcoglycan, Gamma SG, LGMD2C, MAM, SCARMD2, SCG3, SGCG, TYPE | |
SGCG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |