Learn More
SGE™ ConnecTite™ Liners for PerkinElmer™ GC
Designed for PerkinElmer AutoSystem™ and Clarus 500™ GCs
Brand:  SGE™ 092344
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15259033
  • 2950.00 SEK / Pack of 5
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 6.2mm OD, 4mm ID; 92mm length
Specifications
Specifications
Connectite standard liner
0.4cm
9.2cm
5/Pack
0.62cm
Perkin Elmer™ autosystem, clarus 500
Videos