Learn More
SGE™ ConnecTite™ Liners for PerkinElmer™ GC
Designed for PerkinElmer AutoSystem™ and Clarus 500™ GCs
Brand:  SGE™ 092344
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15259033
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 6.2mm OD, 4mm ID; 92mm length
Specifications
Specifications
Connectite standard liner
0.4cm
9.2cm
5/Pack
0.62cm
Perkin Elmer™ autosystem, clarus 500
Videos