Learn More
SGE™ ConnecTite™ Liners for Thermo Scientific™ GC
Designed for use with Thermo Scientific TRACE™ and FOCUS™ GCs
Brand:  SGE Analytical Science 09233501
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15269023
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Specifications
Specifications
Connectite top hole liner
0.5 cm
10.5 cm
1/Pk.
0.8 cm
Thermo Scientific GC Trace and Focus
Videos