Learn More
SGE™ ConnecTite™ Liners for Varian™ GC
For use with Varian 1075/1077 GC models
Brand:  SGE™ 092341
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239033
  • 3814.19 SEK / Pack of 5
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Every batch of inlet liners tested for activity using the EPA 8081B method
  • 6.3mm OD, 4mm ID; 72mm length
Specifications
Specifications
Connnectite bottom hole liner
0.4cm
7.2cm
5/Pack
0.63cm
Varian 1075/1077 GC models
Videos