Learn More
SGE™ ConnecTite™ Liners for Varian™ GC
For use with Varian 1075/1077 GC models
Brand:  SGE™ 09234101
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15249033
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • Every batch of inlet liners tested for activity using the EPA 8081B method
  • 6.3mm OD, 4mm ID; 72mm length
Specifications
Specifications
Connnectite bottom hole liner
0.4cm
7.2cm
1/Pack
0.63cm
Varian 1075/1077 GC models
Videos