Learn More
Trajan™ Diamond MS Manual Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
1215.00 SEK
Specifications
Includes | Removable ultra smooth needle |
---|---|
Product Type | Diamond MS Manual Syringe |
Quantity | Ea. |
Connection Type | Removable |
Product Code | Brand | Volume (Metric) | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Price | Quantity & Availability | ||||
13187966
|
Trajan™
002750 |
10 μL | 10R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13197966
|
Trajan™
003750 |
25 μL | 25R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13107976
|
Trajan™
004750 |
50 μL | 50R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13117976
|
Trajan™
005750 |
100 μL | 100R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13127976
|
Trajan™
006750 |
250 μL | 250R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13137976
|
Trajan™
007750 |
500 μL | 500R-GT-MS1 |
1215.00 SEK
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Removable ultra smooth needle | |
Diamond MS Manual Syringe | |
Ea. | |
Removable |