Learn More
Trajan™ Replacement Needles for Thermo Scientific GC Syringes
Cone tip style. Trajan Scientific and Medical™ Replacement Needles for Thermo Scientific GC Autosampler Syringes are not gas tight.
Brand:  Trajan™ 031535
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15228523
  • 875.00 SEK / Pack of 3
Estimated Shipment: 02-07-2024
to see stock.
Description
Description
  • Needle length is the exposed length of needle
  • Fits Thermo Scientific™ syringes for AS3000, AS2000 and TriPlus RSH Autosamplers
Specifications
Specifications
26 G
0.47 mm
10 μL
Cone
0.15 mm
Replacement Needles for Thermo Scientific GC Syringes
80 mm