Learn More
Trajan™ GC Septa Enduro Blue Plug
GP GC septa is designed for non-demanding routine, low temperature applications
Brand: Trajan™ 041890
Additional Details : Weight : 0.50000kg
Description
- Designed for non-demanding, routine applications
- Reliable seal against carrier gas pressure in inlet
- No bleed or contamination into GC system
- Maximum operating temperature of 275°C
Specifications
Enduro Blue Septa | |
50/Pk. |