missing translation for 'onlineSavingsMsg'
Learn More

SGE™ MEPS™ BINs

Available in needle styles to suit LC and GC applications

Brand:  SGE™ 2900408

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15269233

  • 3695.00 SEK / Pack of 5


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Description

Description

The MEPS approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries.

  • For GC applications: needle is 23 gauge, 0.63mm OD, cone point style
  • For LC applications: needle is 22 gauge, 0.72mm OD, LC point style
  • Typical BIN life is conservatively about 40-100 samples but actual life is dependant on the sample matrix
  • The dimensions of the sorbent bed ensure that the sorbent performance remains identical to conventional SPE devices used for extraction of similar samples
  • Sample preparation for complex biological samples is readily adapted to MEPS and reduces the volume of sample and reagents required for extraction when compared with conventional SPE and other “micro extraction” procedures
  • Needles are packed in a sealed blister pack of 5 needles to ensure their integrity
  • Suitable for use with eVol™ automated syringes

Specifications

Specifications

Silica
0.07cm
22G
MEPS bin
LC
45μm
CatIon Exchange
A300 Plus, Thermo Scientific™, Varian 8400, 100μL CTC Analytics systems
60Å
SCX
MEPS
Videos