Learn More
Trajan™ MEPS BINs for GC Applications
MEPS is Micro Extraction by Packed Sorbent. Provides miniaturization of conventional sample preparation devices
Brand:  Trajan™ 2900103
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15269223
  • 3292.35 SEK / Pack of 5
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
GC
Gas chromatography
23g
MEPS BINs
Cone point needle
45μm
0.63mm
HTA 300APlus, Thermo Scientific, Varian 8400 and 100μL CTC Analytics systems
60Å
C8+SCX
10μL to 1mL or more
Videos