Learn More
Trajan™ MEPS BINs for LC Applications
MEPS is Micro Extraction by Packed Sorbent. Provides miniaturization of conventional sample preparation devices
Brand:  Trajan™ 2900404
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239233
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
LC
Liquid chromatography
22g
MEPS BINs
LC point needle
45μm
0.72mm
HTA 300APlus, Thermo Scientific, Varian 8400 and 100μL CTC Analytics systems
60Å
C2
10μL to 1mL or more
Videos