Learn More
Trajan™ PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
Brand:  Trajan™ 0624351
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15208783
  • 817.35 SEK / Pack of 5
Estimated Shipment
14-12-2020
Add to basket
Description
Description
  • Robust - PEEK polymer exterior and fused silica inner tubing
  • Smooth wall surface giving excellent flow characteristics
  • Compatible with most organic solvents
  • Inertness - extremely low absorption characteristics
  • Precise ID
Specifications
Specifications
50 mm
Tubing
1.5 mm
0.175 mm