Learn More
Trajan™ Replacement Needles for 5μL eVol™ and eVol™ NMR Syringes
Cone tipped replacement needle for use with 5uL NMR Syringes.
Brand: Trajan™ 036912
Additional Details : Weight : 0.20000kg
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, and outer diameter
Specifications
Stainless Steel | |
0.0066 in. | |
0.17 mm | |
2 in. | |
51 mm | |
eVol NMR Syringe (5μL) |
22 G | |
0.02 in. | |
0.72 mm | |
Replacement Needle | |
LC |