Learn More
SGE™ SilFlow™ GC 4 Port Splitter
Highly efficient and reliable micro fluidic platform to improve GC connectivity
Brand:  SGE™ 123735
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15289183
  • 5745.00 SEK / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
  • Does not include 1.1mm OD tubing
Port A: Variable see product descriptions
Port B: 0.25/0.32mm ID
Port C: 0.25/0.32mm ID
Port D: 0.25/0.32mm ID
Specifications
Specifications
Port Splitter
0.25/0.32 mm
SilFlow microchannel device (MCD)
SGE Silflow Microchannel Device