Learn More
Trajan™ SilTite™ FingerTite Agilent INJ Base Seal
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073640
Additional Details : Weight : 0.01000kg
Product Code. 15268983
  • 680.00 SEK / Pack of 2
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Seal
2/Pk.
Agilent INJ