Learn More
Trajan™ SilTite™ FingerTite Agilent INJ Base Seal
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan™ 073640
Additional Details : Weight : 0.01000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Seal | |
2/Pk. | |
Agilent INJ |