missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23394-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to SGK1.
Serum and glucocorticoid-regulated protein kinase 1 (SGK1) is a member of the AGC family of serine/threonine kinases that plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. SGK1 functions as a regulator of cell survival and ion channels and transporters such as ENaC or KCNA3/Kv1.3. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.Specifications
SGK1 | |
Polyclonal | |
Unconjugated | |
SGK1 | |
SGK, SGK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20393, 6446 | |
-20°C | |
Liquid |
Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00141, Q9WVC6 | |
SGK1 | |
SGK1 Fusion Protein Ag17888 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |