missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12226-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described.Specifications
SH2B1 | |
Polyclonal | |
Unconjugated | |
SH2B1 | |
KIAA1299, PSM, SH2B, SH2B adapter protein 1, SH2B adaptor protein 1, SH2B1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25970 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRF2 | |
SH2B1 | |
SH2B1 Fusion Protein Ag2863 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |