Learn More
Abnova™ SH2D2A Recombinant Protein
4055.00 SEK - 6150.00 SEK
Specifications
Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 68.42
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16119805
View Documents Promotion Details
Abnova™
H00009047-P01.10UG
10 ug
4055.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-07-2022
to see stock.
16129805
View Documents Promotion Details
Abnova™
H00009047-P01.25UG
25 ug
6150.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-07-2022
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
Human
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
68.42
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Solution
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
SH2D2A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.