missing translation for 'onlineSavingsMsg'
Learn More

SH2D4A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15957-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895364

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
Specifications

Specifications

SH2D4A
Polyclonal
Unconjugated
SH2D4A
Protein SH(2)A, SH2 domain containing 4A, SH2A, SH2D4A
Rabbit
Antigen Affinity Chromatography
RUO
63898
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H788
SH2D4A
SH2D4A Fusion Protein Ag8713
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers