missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D4A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15957-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.Specifications
SH2D4A | |
Polyclonal | |
Unconjugated | |
SH2D4A | |
Protein SH(2)A, SH2 domain containing 4A, SH2A, SH2D4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63898 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H788 | |
SH2D4A | |
SH2D4A Fusion Protein Ag8713 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |