missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539452
Additional Details : Weight : 0.00900kg
Specifications
SH2D5 | |
Polyclonal | |
Unconjugated | |
SH2D5 | |
SH2D5 | |
A synthetic peptide derived from the C-terminal region of human SH2D5 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6ZV89, Q8JZW5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230863, 366489, 400745 | |
-20°C | |
Liquid |