missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55320-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
SH2D5 | |
Polyclonal | |
Unconjugated | |
SH2D5 | |
SH2 domain containing 5, SH2D5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230863, 400745 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZV89, Q8JZW5 | |
SH2D5 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |