missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BGR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SH3BGR |
---|---|
Concentration | 0.54 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850775
|
Proteintech
21161-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840775
|
Proteintech
21161-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SH3BGR is a protein coding gene.Specifications
SH3BGR | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P55822, Q9WUZ7 | |
50795, 6450 | |
SH3BGR Fusion Protein Ag14739 | |
Primary | |
-20°C | |
SH3BGR |
0.54 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
21 GARP, 21 glutamic acid rich protein, SH3BGR, SH3BGR protein | |
Sh3bgr | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |