missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3D19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21499-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May play a role in regulating A disintegrin and metalloproteases (ADAMs) in the signaling of EGFR-ligand shedding. May be involved in suppression of Ras-induced cellular transformation and Ras-mediated activation of ELK1. Plays a role in the regulation of cell morphology and cytoskeletal organization.Specifications
SH3D19 | |
Polyclonal | |
Unconjugated | |
SH3D19 | |
ADAM binding protein Eve 1, EBP, EEN binding protein, EVE1, Kryn, SH3 domain containing 19, SH3D19, SH3P19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
152503, 27059, 295171 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5HYK7, Q91X43 | |
SH3D19 | |
SH3D19 Fusion Protein Ag15915 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |