Learn More
Epredia™ Cryochrome™ Embedding Resin
Product Code. 12570017 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Pink
Cryochrome Sampler Colors
Cryochrome Yellow
Unit Size:
120mL
Each
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12570017 Cryochrome Sampler Colors Pack of 4
12533527 Cryochrome Blue Pack of 4
12560017 Cryochrome Yellow Pack of 4
12726087 Cryochrome Sampler Colors Each
12543527 Cryochrome Green 120mL
12580017 Cryochrome Pink Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12570017 Supplier Epredia™ Supplier No. 9990426
  • ONLINE EXCLUSIVE PRICE
    1185.00 SEK
    1022.06 SEK / Paket med 4
    Save 162.94 SEK
    13% Av
 Restricted Item
I lager
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryochrome Sampler Colors
Quantity 120mL