missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHCBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12672-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the larger subunits of TFIID that is stably associated with the TFIID complex. It contributes to interactions at and downstream of the transcription initiation site, interactions that help determine transcription complex response to activators.Specifications
SHCBP1 | |
Polyclonal | |
Unconjugated | |
SHCBP1 | |
PAL, SHCBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20419, 79801 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NEM2, Q9Z179 | |
SHCBP1 | |
SHCBP1 Fusion Protein Ag3355 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |