missing translation for 'onlineSavingsMsg'
Läs mer

PLAU, Sheep, Polyclonal Antibody, Abnova™

Produktkod. 16091337
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mL
Förpackningsstorlek:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16091337 1 mL 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16091337 Leverantör Abnova Leverantörsnummer PAB14929.1mL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Sheep polyclonal antibody raised against native PLAU.

This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifikationer

Antigen PLAU
Användningsområden ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Sheep polyclonal antibody raised against native PLAU.
Utspädning The optimal working dilution should be determined by the end user.
Formulering In serum
Gen PLAU
Gene Alias ATF/UPA/URK/UROKINASE/u-PA
Gensymboler PLAU
Värd art Sheep
Immunogen Native purified two chain human urinary type high molecular weight PLAU from pooled urine.
Kvantitet 1 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 5328
Testspecificitet This antibody reacts only with human u-PA isolated from kidney cell culture tissue. No reactivity with other human proteins. Identity established by double diffusion with u-PA isolated from human kidney tissue and E. coli-derived recombinant u-PA. No cross reactivity with human tissue type plasminogen activator.
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.