Learn More
Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with shelf kits designed for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand: Thermo Scientific™ SK500
2859.47 SEK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Maximize storage space with Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers; designed for use with with Thermo Scientific™ Forma™ 8800, HERAfreeze™ HFU-T, Revco™ UxF, and TSU Series freezers.
Specifications
Includes one shelf and clips | |
Shelf Kit | |
1 | |
Forma 88500; TSU500; HFU500T; Revco UxF500 |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Thermo Scientific Revco UxF freezers
-
Thermo Scientific Revco UxF freezers
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
50Hours
8Minutes
15Seconds
41 -
Multiple Manufacturers
Illuminating Protein Pathways
Shining a light on the tools that drive protein discovery.Promo Code: 24148Expires: 2025-08-22
New Offer Save Now