Läs mer
Shikimate-3-phosphate Trisodium Salt (~90%), TRC
Produktkod. 30133474
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133474 5 mg
30007450 10 mg
30010838 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133474 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS3570265MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 143393-03-7
Molekylformel C7 H8 O8 P . 3 Na
Molekylvikt (g/mol) 320.08
Synonym (3R,4S,5R)-4,5-Dihydroxy-3-(phosphonooxy)-1-cyclohexene-1-carboxylic Acid Sodium Salt, [3R-(3α,4α,5β)]-4,5-Dihydroxy-3-(phosphonooxy)-1-cyclohexene-1-carboxylic Acid Trisodium Salt, S-3-P Trisodium
IUPAC-namn trisodium;(3R,4S,5R)-4,5-dihydroxy-3-phosphonatooxycyclohexene-1-carboxylate
LEDER [Na+].[Na+].[Na+].O[C@@H]1CC(=C[C@@H](OP(=O)([O-])[O-])[C@H]1O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material Shikimate-3-phosphate Trisodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C7H11O8P.3Na/c8-4-1-3(7(10)11)2-5(6(4)9)15-16(12,13)14;;;/h2,4-6,8-9H,1H2,(H,10,11)(H2,12,13,14);;;/q;3*+1/p-3/t4-,5-,6+;;;/m1.../s1
Formel vikt 319.965
Procent renhet >95%