missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Shirt with Long sleeve 7482-Black
Click to view available options
Size:
3X-Large/4X-Large
5X-Large/6X-Large
Medium/Large
Small
X-Large/2X-Large
Description
- Longsleeve men uvex suXXeed
- Color: Black Material: 60% Dryarn, 28% wool , 8% PA, 4% EL, 250 gsm
- Seamless underwear, without annoying seams, moisture-regulating, with optimal fit for maximum comfort, soft cuffs, merino wool for best insulation, odor neutralizing
- Size: S- 5XL/6XL
- Care: 30 degree washing, no chlorine, no tumble drying, no iron, no chemical laundry
Specifications
Size | Medium/Large |
Color | Black |
Thickness (Metric) | 250 g/m² |