missing translation for 'onlineSavingsMsg'
Learn More

SHISA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22197-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891725

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SHISA4 plays an essential role in the maturation of presomitic mesoderm cells by individual attenuation of both FGF and WNT signaling. The Shisa family of single-transmembrane proteins is characterized by an N-terminal cysteine-rich domain and a proline-rich C-terminal region. Its founding member, Xenopus Shisa, promotes head development by antagonizing Wnt and FGF signaling. Shisa physically interacted with immature forms of the Wnt receptor Frizzled and the FGF receptor within the ER and inhibited their posttranslational maturation and trafficking to the cell surface. Loss of Shisa function sensitized the neuroectoderm to Wnt signaling and suppressed head formation during gastrulation.
Specifications

Specifications

SHISA4
Polyclonal
Unconjugated
SHISA4
C1orf40, hShisa4, Protein shisa 4, SHISA4, TMEM58, Transmembrane protein 58
Rabbit
Antigen Affinity Chromatography
RUO
149345, 77552
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CA71, Q96DD7
SHISA4
SHISA4 Fusion Protein Ag17357
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers