Learn More
SHKBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559773
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PVFPVKRRNR HSLVGPQQLG GRPAPVRRSN TMPPNLGNAG LLGRMLDEKT PPSPSGQPEE PGMVRLVCGH HNWIAVAYTQ FLVCYRLKEA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
Inhibits CBL-SH3KBP1 complex mediated down-regulation of EGFR signaling by sequestration of SH3KBP1. Binds to SH3KBP1 and prevents its interaction with CBL and inhibits translocation of SH3KBP1 to EGFR containing vesicles upon EGF stimulation.Specifications
SHKBP1 | |
Polyclonal | |
Unconjugated | |
SHKBP1 | |
B930062H15Rik; PP203; RGD1309281; Sb1; SETA binding protein 1; SETA-binding protein 1; SH3KBP1 binding protein 1; SH3KBP1-binding protein 1; Shkbp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TBC3 | |
SHKBP1 | |
Recombinant protein corresponding to Human SHKBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only