Läs mer
SHP099, MedChemExpress
Produktkod. 16474296
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16474296 100 mg
16435727 5 mg
16419556 50 mg
16412988 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16474296 Leverantör Medchem Express Leverantörsnummer HY100388
MedChemExpress SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC50 of 70 nM.
SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC50 of 70 nM.
Specifikationer
Kemiskt namn eller material SHP099
CAS 1801747-42-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C16H19Cl2N5
Kvantitet 100 mg
Löslighetsinformation DMSO : 12 mg/mL (34.07 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.84 mM; Need ultrasonic)
LEDER NC1=NC(N2CCC(C)(N)CC2)=CN=C1C3=CC=CC(Cl)=C3Cl
Molekylvikt (g/mol) 352.26
Formel vikt 352.26
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease