Learn More
SIAH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563880
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SFHPHHLSHH HCHHRHHHHL RHHAHPHHLH HQEAGLHANP VTPCLCMCPL FSCQWEGRLE VVVPHLRQIH RVDILQGAEI VFLATDMHLP APADWIIMHS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
SIAH3 Polyclonal antibody specifically detects SIAH3 in Human samples. It is validated for Immunohistochemistry.Specifications
SIAH3 | |
Polyclonal | |
Unconjugated | |
SIAH3 | |
seven in absentia homolog 3; siah E3 ubiquitin protein ligase family member 3; SIAH3; siah-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283514 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IW03 | |
SIAH3 | |
Recombinant protein corresponding to Human SIAH3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only