Learn More
Bohlender™ SICCO™ Big Star Desiccator Protect
Big Star Desiccator Protect Orange made of acrylic glass and aluminum frame.
Brand: Bohlender™ V1926-07
Additional Details : Weight : 150.00000kg
Description
- Aluminum frame, orange panels made of acrylic glass to reduce light incidence and protect against ultraviolet radiation
- Includes two shelves made of stainless steel, tray and hygrometer
- Usable with up to 8 shelves
- Orange panels reduce light incidence
- For storage of light-sensitive substances
- Door with magnetic catch and circular rubber seal
- Variable height shelves
- Easy to read electronic hygrometer
- Telescopic shelves
Specifications
Orange | |
41 gal. | |
22 in. | |
22 in. | |
23 in. | |
40 lb. |
Acrylic | |
156 L | |
560 mm | |
560 mm | |
580 mm | |
18 kg |