Läs mer
SID 3712249, MedChemExpress
Produktkod. 16487868
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487868 10 mg
16483676 25 mg
16463817 50 mg
16447686 100 mg
16420338 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16487868 Leverantör Medchem Express Leverantörsnummer HY19731
MedChemExpress SID 3712249 (MiR-544 Inhibitor 1) is an inhibitor of the biogenesis of microRNA-544 (miR-544).
SID 3712249 (MiR-544 Inhibitor 1) is an inhibitor of the biogenesis of microRNA-544 (miR-544).
Specifikationer
Kemiskt namn eller material SID 3712249
CAS 522606-67-3
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C17H21N7
Kvantitet 10 mg
Synonym MiR-544 Inhibitor 1
Löslighetsinformation DMSO : 16.67 mg/mL (51.55 mM; Need ultrasonic)
LEDER NC1=C2C(C=C(N3CCCC3)N=C2N)=C(C#N)C(N4CCCC4)=N1
Molekylvikt (g/mol) 323.4
Formel vikt 323.4
Procent renhet 98.35%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death