Learn More
Invitrogen™ Human SIK3, GST Tag Recombinant Protein
SIK3 (QSK) is a serine/threonine-protein kinase, belongs to QIK subfamily. Recombinant human SIK3 (1-307) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
4230.00 SEK - 64700.00 SEK
Specifications
Gene Alias | QSK, SIK-3, L19, FLJ12240, KIAA0999 |
---|---|
Protein Length | aa 59-365 |
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15409033
|
Invitrogen™
PV6403 |
10μg |
4230.00 SEK
10µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15429033
|
Invitrogen™
PV6405 |
1mg |
64700.00 SEK
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15419033
|
Invitrogen™
PV6404 |
100μg |
14360.00 SEK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The phosphorylation of SIK3 by LKB1 through the 14-3-3 binding enhances its catalytic activity and leads its localization to punctate structures within the cytoplasm. Overexpression of SIK3 promotes G1/S cell cycle progression with ovarian cancer. There are two sites (H331L and A1103V) were mutated at significant frequency in breast cancer. SIK3 is a novel tumor-associated antigen (TAA).
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
QSK, SIK-3, L19, FLJ12240, KIAA0999 | |
Human | |
GST Tag |
aa 59-365 | |
Recombinant |