Läs mer
Silatrane, TRC
Produktkod. 30425788
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425788 1 g
30410132 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425788 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS4330001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 283-60-3
Molekylformel C6 H13 N O3 Si
Molekylvikt (g/mol) 175.26
Synonym 2,8,9-Trioxa-5-aza-1-silabicyclo[3.3.3]undecane, NSC 626544
IUPAC-namn 4,6,11-trioxa-1-aza-5-silabicyclo[3.3.3]undecane
LEDER C1CN2CCO[SiH](OCC2)O1
Specifikationer
Kemiskt namn eller material Silatrane
Kvantitet 1 g
InChI formel InChI=1S/C6H13NO3Si/c1-4-8-11-9-5-2-7(1)3-6-10-11/h11H,1-6H2
Formel vikt 175.0665