Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand: BRAND™ 27012
Additional Details : Weight : 0.16400kg
Description
Specifications
To Deliver | |
B | |
Upper end cotton plug | |
2 x Yellow | |
360 mm |
DIN EN ISO 835 | |
0.1 mL | |
6 | |
20 mL |