Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
PEEKsil tubing™ is a mechanically robust polymer sheathed fused silica tubing for use in HPLC systems
Brand: Trajan Scientific and Medical™ 0624269
Additional Details : Weight : 0.01000kg
Description
- Robust - PEEK polymer exterior and fused silica inner tubing
- Smooth wall surface giving excellent flow characteristics
- Compatible with most organic solvents
- Inertness - extremely low absorption characteristics
- Precise ID
Specifications
Tubing | |
Direct Replacement for Conventional Stainless Steel As Well As a Replacement For Peek Tubing Used in Liquid Chromatography Systems | |
5 μm | |
500 mm | |
1380 bar |
Natural | |
1/32 in. | |
500 mm | |
Silica |