Läs mer
Sildenafil Dimer Impurity, TRC
Produktkod. 17903065
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17903065 100 mg
17902965 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17903065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS435020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346602-67-2
Molekylformel C38 H46 N10 O8 S2
Molekylvikt (g/mol) 834.96
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5,5'-[1,4-piperazinediylbis[sulfonyl(6-ethoxy-3,1-phenylene)]]bis[1,6-dihydro-1-methyl-3-propyl-, Sildenafil Dimer (Bis-1,4-[4-ethoxy-3-(6,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonyl]piperazine)
IUPAC-namn 5-[2-ethoxy-5-[4-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]sulfonylpiperazin-1-yl]sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CC4)S(=O)(=O)c5ccc(OCC)c(c5)C6=Nc7c(CCC)nn(C)c7C(=O)N6
Specifikationer
Kemiskt namn eller material Sildenafil Dimer
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C38H46N10O8S2/c1-7-11-27-31-33(45(5)43-27)37(49)41-35(39-31)25-21-23(13-15-29(25)55-9-3)57(51,52)47-17-19-48(20-18-47)58(53,54)24-14-16-30(56-10-4)26(22-24)36-40-32-28(12-8-2)44-46(6)34(32)38(50)42-36/h13-16,21-22H,7-12,17-20H2,1-6H3,(H,39,41,49)(H,40,42,50)
Formel vikt 834.2941
Procent renhet >95%