missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ SilFlow™ Ferrules
Microfluidic platform for multi-dimensional GC.
1360.00 SEK - 2360.00 SEK
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
15259173
|
Trajan Scientific and Medical™
123713 |
SilFlow Ferrules 0.35 mm ID | 5/Pk. |
2360.00 SEK
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
15269173
|
Trajan Scientific and Medical™
123715 |
SilFlow Blanking Ferrules and Pins | 10/Pk. |
1360.00 SEK
Pack of 5 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
15299183
|
Trajan Scientific and Medical™
123742 |
SilFlow Ferrules 0.8 mm ID | 10/Pk. |
2360.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
Description
- Enhanced GC connectivity
- Replacement ferrules and tubing available
Specifications
Ferrules and pins |