Learn More
Thermo Scientific Alfa Aesar 1GR Silica gel, HPLC grade, spherical, 2.2 micron APS, 120 angstroms, 99.99+% 1g
Brand: Thermo Scientific Alfa Aesar 045798.03
1GR Silica gel, HPLC grade, spherical, 2.2 micron APS, 120 angstroms, 99.99+% 1g
Additional Details : CAS Number : 63231-67-4 Weight : 0.00100kg