missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, HPLC/UHPLC grade, spherical, 1.8 micron APS, 110 angstroms, 99.99+%, Thermo Scientific™

Catalog Number p-7059954
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Spherical, S.A. 340m
Material Silica Gel
Quantity 1 g
Particle Size 1.8 Micron APS, 110Å
Percent Purity ≥99.99%
Grade HPLC
Surface Area 340m2/g
Chemical Name or Material Silica gel, P.V. 0.95cc/g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'