missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, preparative chromatography grade, spherical, 50 micron APS, 60 angstroms, 99.9+%, Thermo Scientific Chemicals

Catalog Number p-7059973
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O

Silica gel was used to compare MCM-41 type mesoporous adsorbents. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. It is a useful product for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Spherical, S.A. 650m
Material Silica Gel
Quantity 25 g
Particle Size 50 Micron APS, 60Å
Solubility Information Insoluble in water. Soluble in hydrofluoric acid.
Percent Purity ≥99.9%
Grade Preparative Chromatography
Surface Area 650m2/g
Chemical Name or Material Silica gel, P.V. 1.0cc/g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'