Learn More
Thermo Scientific Alfa Aesar 25GR Silica gel, preparative chromatography grade, spherical, 7.5 micron APS, 80 angstroms, 99.99+%
Brand: Thermo Scientific Alfa Aesar 045815.14
25GR Silica gel, preparative chromatography grade, spherical, 7.5 micron APS, 80 angstroms, 99.99+%
Additional Details : CAS Number : 63231-67-4 Weight : 0.02500kg