missing translation for 'onlineSavingsMsg'
Läs mer

Silicon Tetrachloride, Sigma-Aldrich™

Product Code. 15854221
Change view
Click to view available options
Kvantitet:
100 mL
1 L
Unit Size:
1 L
100ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
15854221 1 L 1 L
15844221 100 mL 100ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15854221 Supplier Sigma Aldrich Supplier No. 8077061000
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-04-7

Specifications

Kokpunkt 57°C/1013 hPa
Kemiskt namn eller material Silicon Tetrachloride
Smältpunkt -70°C
Kvantitet 1 L
Densitet 1.48 g/cm3 at 20°C
MDL-nummer MFCD00011229
Synonym Silicon tetrachloride, Silicon(IV) chloride, Tetrachlorosilane, Silicon tetrachloride
Molekylvikt (g/mol) 169.9
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.