Läs mer
Borer Chemie™ Silicone-free Grease Tube
Produktkod. 10404681
Change view
Produkttyp:
Standard
Förpackningsstorlek:
60g
Produktkod. Produkttyp Förpackningsstorlek
10404681 Standard 60g
1 options
Produktkod. 10404681 Leverantör Borer Chemie™ Leverantörsnummer 514215.00CA15
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Standard quality for desiccators, ground joints, stopcocks and apparatuses. Borer Chemie™ Silicone-Free Grease Tubes are suitable for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metal.
  • Excellent thermal stability
  • Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
  • Constant lubricating consistency between -40°C and +250°C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Insoluble in water and low molecular mono- and polyhydric alcohols
Specifikationer
Produkttyp Standard
Kvantitet 60 g