Learn More
Silicone Oil, For oil baths up to 250°C, MilliporeSigma™
Product Code. 15885030
Change view
Click to view available options
Quantity:
1000 mL
Packaging:
Plastic Bottle
Unit Size:
1 L
Product Code. Quantity Packaging Unit Size
15885030 1000 mL Plastic Bottle 1 L
1 options
Product Code. 15885030 Supplier Supelco Supplier No. 1077421000
 Restricted Item
Beräknad leverans: 18-11-2026
Specifications
Boiling Point >250°C (1013hPa)
CAS 68083-14-7
Color Colorless
Physical Form Liquid
Quantity 1000 mL
Molecular Formula C16H22O2Si2
Flash Point 315°C
Synonym diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.
InChI Key ARWRSWALIGRKQA-UHFFFAOYSA-N
SMILES CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2
IUPAC Name methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane
Molecular Weight (g/mol) 302.52
PubChem CID 121233058
Packaging Plastic Bottle
Vapor Pressure 47 hPa (20°C)
Density 1.04g/3 (20°C)
Chemical Name or Material Sillicone