Learn More
Fisherbrand™ Silikonslang
Product Code. 11507083 Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Diameter (metrisk) Inre:
1,0 mm
2 mm
3 mm
4 mm
5 mm
6 mm
6,5 mm
8 mm
9 mm
10,0 mm
12,5 mm
16 mm
Tjocklek (metrisk):
0,75 mm
1,0 mm
1,5 mm
2 mm
2,0 mm
2,25 mm
25 mm
missing translation for 'unitSize'
10m
Product Code. 11507083
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
För konsekvent pålitliga och exakta vätskeöverföringar.
Available in different diameters
Specifications
Diameter (metrisk) Inre 6 mm
Total längd (metrisk) 10 m
Tjocklek (metrisk) 1,5 mm